Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9B2
Entrez Gene ID
711024
Gene Name
solute carrier family 9 member B2
Gene Aliases
NHEDC2
Chromosome Location
Chr.5: 101438762 - 101490227 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001110438.4 XP_001110438.1 - 845 129
XM_015138894.2 XP_014994380.2 - 845 129

View Full Product Details