Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA13
Entrez Gene ID
703583
Gene Name
carbonic anhydrase 13
Gene Aliases
-
Chromosome Location
Chr.8: 85791958 - 85828740 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095487.4 XP_001095487.1 - 734 131
GenBank mRNA JV650826.1 - - 216 131

View Full Product Details