Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSX1
Entrez Gene ID
722771
Gene Name
msh homeobox 1
Gene Aliases
-
Chromosome Location
Chr.5: 8045991 - 8049871 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040416.2 NP_001035506.1 - 716 123

View Full Product Details