Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM21
Entrez Gene ID
715640
Gene Name
ADAM metallopeptidase domain 21
Gene Aliases
-
Chromosome Location
Chr.7: 132536314 - 132566157 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194322.3 NP_001181251.3 - 1117 102
XM_028850521.1 XP_028706354.1 - 2097 102
XM_028850522.1 XP_028706355.1 - 1634 102
XM_028850523.1 XP_028706356.1 - 2347 102
XM_028850524.1 XP_028706357.1 - 1621 102
XM_028850525.1 XP_028706358.1 - 1475 102
XM_028850526.1 XP_028706359.1 - 1950 102
XM_028850528.1 XP_028706361.1 - 1474 102
XM_028850529.1 XP_028706362.1 - 1301 102

View Full Product Details