Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLRP6
Entrez Gene ID
720741
Gene Name
NLR family pyrin domain containing 6
Gene Aliases
-
Chromosome Location
Chr.14: 270568 - 278431 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114114.1 NP_001107586.1 - 27 77
GenBank mRNA EU338447.1 - - 27 77

View Full Product Details