Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPH2
Entrez Gene ID
703254
Gene Name
heterogeneous nuclear ribonucleoprotein H2
Gene Aliases
HNRPH2
Chromosome Location
Chr.X: 98221501 - 98227705 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193993.2 NP_001180922.1 - 113 67

View Full Product Details