Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLYATL3
Entrez Gene ID
708571
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
GLYATL2
Chromosome Location
Chr.4: 119965306 - 119979244 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194067.2 NP_001180996.2 - 346 80

View Full Product Details