Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED1
Entrez Gene ID
713618
Gene Name
transmembrane p24 trafficking protein 1
Gene Aliases
EGK_10111
Chromosome Location
Chr.19: 10593549 - 10597389 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194250.2 NP_001181179.1 - 298 72
GenBank mRNA JU327678.1 - - 119 72
JU479250.1 - - 122 72
JV046038.1 - - 203 72

View Full Product Details