Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRGX
Entrez Gene ID
719355
Gene Name
dorsal root ganglia homeobox
Gene Aliases
-
Chromosome Location
Chr.9: 47982075 - 48014029 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194598.1 NP_001181527.1 - 244 69
XM_028853539.1 XP_028709372.1 - 329 69

View Full Product Details