Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRGX
Entrez Gene ID
719355
Gene Name
dorsal root ganglia homeobox
Gene Aliases
-
Chromosome Location
Chr.9: 47982075 - 48014029 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194598.1 NP_001181527.1 - 427 69
XM_028853539.1 XP_028709372.1 - 512 69

View Full Product Details