Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNK17
Entrez Gene ID
719574
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
-
Chromosome Location
Chr.4: 130065069 - 130080452 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194723.2 NP_001181652.2 - 510 72
XM_015136266.2 XP_014991752.2 - 796 72

View Full Product Details