Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHEJ1
Entrez Gene ID
701542
Gene Name
non-homologous end joining factor 1
Gene Aliases
-
Chromosome Location
Chr.12: 106511047 - 106600352 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245955.1 NP_001232884.1 - 176 110
XM_015110986.2 XP_014966472.1 - 323 110
XM_028830469.1 XP_028686302.1 - 508 110
GenBank mRNA HM486750.1 - - 176 110
JU477250.1 - - 274 110
JV636380.1 - - 274 110

View Full Product Details