Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIMD2
Entrez Gene ID
717889
Gene Name
LIM domain containing 2
Gene Aliases
EGK_08863
Chromosome Location
Chr.16: 60149302 - 60153814 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260589.1 NP_001247518.1 - 187 91
XM_028835761.1 XP_028691594.1 - 492 91
XM_028835762.1 XP_028691595.1 - 333 91
XM_028835763.1 XP_028691596.1 - 192 91
XM_028835764.1 XP_028691597.1 - 1518 91
GenBank mRNA JU336515.1 - - 212 91
JU477326.1 - - 187 91
JV629726.1 - - 139 91

View Full Product Details