Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUSC1
Entrez Gene ID
708101
Gene Name
tumor suppressor candidate 1
Gene Aliases
-
Chromosome Location
Chr.15: 58051458 - 58052780 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265623.1 NP_001252552.1 - 322 63
GenBank mRNA JV614130.1 - - 328 63
JV636108.1 - - 322 63

View Full Product Details