Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR34A
Entrez Gene ID
100315136
Gene Name
microRNA mir-34a
Gene Aliases
mir-34a, mml-mir-34a
Chromosome Location
Chr.1: 215342661 - 215342770 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032030.1 - 61 80

View Full Product Details