The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022223.2 | NP_071559.2 | 3 - 4 | 404 | 135 |
GenBank mRNA | AB008908.1 | - | - | 419 | 135 |
AB079500.1 | - | 3 - 4 | 404 | 135 | |
AF348523.1 | - | - | 298 | 135 |