Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crabp1
Entrez Gene ID
25061
Gene Name
cellular retinoic acid binding protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 59344097 - 59352135 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105716.1 NP_001099186.1 3 - 4 464 87

View Full Product Details