Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3rf2
Entrez Gene ID
307472
Gene Name
SH3 domain containing ring finger 2
Gene Aliases
Posh2, Ppp1r39
Chromosome Location
Chr.18: 36371041 - 36471020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034187.1 NP_001029359.1 8 - 9 2082 82
GenBank mRNA BC100155.1 - 8 - 9 2082 82

View Full Product Details