Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mras
- Entrez Gene ID
-
25482
- Gene Name
- muscle RAS oncogene homolog
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 107629028 - 107656851 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D89863.1 |
- |
- |
417 |
62 |
View Full Product Details