The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013019.2 | NP_037151.2 | 4 - 5 | 411 | 78 |
XM_008772599.1 | XP_008770821.1 | 4 - 5 | 1250 | 78 | |
GenBank mRNA | BC062016.1 | - | 4 - 5 | 411 | 78 |
X06890.1 | - | 4 - 5 | 343 | 78 |