The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013027.3 | NP_037159.4 | 1 - 2 | 115 | 62 |
GenBank mRNA | BC087625.1 | - | 1 - 2 | 91 | 62 |
DN931873.1 | - | 1 - 2 | 137 | 62 | |
FQ212381.1 | - | 1 - 2 | 101 | 62 | |
FQ212407.1 | - | 1 - 2 | 95 | 62 | |
FQ215130.1 | - | 1 - 2 | 94 | 62 | |
FQ215391.1 | - | 1 - 2 | 96 | 62 | |
FQ216964.1 | - | 1 - 2 | 91 | 62 | |
FQ217212.1 | - | 1 - 2 | 96 | 62 | |
FQ217426.1 | - | 1 - 2 | 97 | 62 | |
FQ217557.1 | - | 1 - 2 | 97 | 62 | |
FQ217567.1 | - | 1 - 2 | 97 | 62 | |
FQ217929.1 | - | 1 - 2 | 97 | 62 | |
FQ221681.1 | - | 1 - 2 | 97 | 62 | |
FQ223747.1 | - | 1 - 2 | 97 | 62 | |
FQ224556.1 | - | 1 - 2 | 101 | 62 | |
FQ224570.1 | - | 1 - 2 | 97 | 62 | |
FQ224606.1 | - | 1 - 2 | 94 | 62 | |
U25264.1 | - | 1 - 2 | 82 | 62 |