Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asns
Entrez Gene ID
25612
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
-
Chromosome Location
Chr.4: 33742876 - 33761100 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013079.2 NP_037211.2 2 - 3 431 80
XM_006236089.2 XP_006236151.1 2 - 3 424 80
GenBank mRNA BC081719.1 - 2 - 3 431 80
U07201.1 - 2 - 3 375 80
U07202.1 - 2 - 3 375 80

View Full Product Details