The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013085.3 | NP_037217.3 | - | 1183 | 72 |
GenBank mRNA | BC105859.1 | - | - | 1183 | 72 |
X65651.1 | - | 10 - 11 | 1214 | 72 |