The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016997.2 | NP_058693.2 | 1 - 2 | 89 | 61 |
GenBank mRNA | X16054.1 | - | 1 - 2 | 85 | 61 |