The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017046.1 | NP_058742.1 | 7 - 8 | 1273 | 76 |
GenBank mRNA | EU627517.1 | - | 6 - 7 | 1175 | 76 |
U37539.1 | - | 7 - 8 | 1227 | 76 | |
U37540.1 | - | 7 - 8 | 1227 | 76 | |
X77933.1 | - | 7 - 8 | 1273 | 76 | |
X78034.1 | - | 7 - 8 | 1256 | 76 |