The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017181.2 | NP_058877.1 | 1 - 2 | 181 | 64 |
GenBank mRNA | BC076381.1 | - | 1 - 2 | 181 | 64 |
FQ219602.1 | - | 1 - 2 | 139 | 64 | |
M77694.1 | - | 1 - 2 | 103 | 64 |