The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033898.1 | NP_001029070.1 | 5 - 6 | 688 | 102 |
NM_022535.2 | NP_071980.2 | 5 - 6 | 688 | 102 | |
GenBank mRNA | AJ001929.1 | - | 5 - 6 | 721 | 102 |
BC101908.1 | - | 5 - 6 | 688 | 102 | |
FQ222101.1 | - | 4 - 5 | 531 | 102 |