The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030826.3 | NP_110453.3 | 1 - 2 | 310 | 77 |
GenBank mRNA | BC058438.1 | - | 1 - 2 | 282 | 77 |
BI281978.1 | - | 1 - 2 | 77 | ||
CB579218.1 | - | 1 - 2 | 310 | 77 | |
FQ211052.1 | - | 1 - 2 | 307 | 77 | |
FQ227523.1 | - | 1 - 2 | 339 | 77 | |
M21210.1 | - | 1 - 2 | 316 | 77 |