Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map1a
Entrez Gene ID
25152
Gene Name
microtubule-associated protein 1A
Gene Aliases
Mtap1a
Chromosome Location
Chr.3: 113251864 - 113272193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030995.1 NP_112257.1 4 - 5 8402 78
XM_006234826.2 XP_006234888.1 5 - 6 8772 78
XM_006234828.2 XP_006234890.1 4 - 5 8676 78
XM_008762145.1 XP_008760367.1 4 - 5 8412 78
GenBank mRNA M83196.1 - 4 - 5 8402 78

View Full Product Details