The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031026.1 | NP_112288.1 | 9 - 10 | 1108 | 82 |
XM_006255054.2 | XP_006255116.1 | 9 - 10 | 1322 | 82 | |
GenBank mRNA | AB008522.1 | - | 4 - 5 | 374 | 82 |
AB008523.1 | - | 4 - 5 | 374 | 82 | |
U15138.1 | - | 9 - 10 | 1108 | 82 |