Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ptafr
- Entrez Gene ID
-
58949
- Gene Name
- platelet-activating factor receptor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 150746294 - 150775675 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U04740.1 |
- |
1 - 2 |
74 |
91 |
View Full Product Details