Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
85426
Gene Name
solute carrier family 5 (sodium/choline cotransporter), member 7
Gene Aliases
Cht1
Chromosome Location
Chr.9: 5293381 - 5330976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053521.1 NP_445973.1 6 - 7 964 93
XM_006244251.2 XP_006244313.1 6 - 7 1067 93
XM_006244253.2 XP_006244315.1 4 - 5 611 93
XM_008766774.1 XP_008764996.1 6 - 7 1115 93
XM_008766775.1 XP_008764997.1 6 - 7 1064 93
GenBank mRNA AB030947.1 - 6 - 7 964 93

View Full Product Details