The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145672.4 | NP_663705.1 | 2 - 3 | 285 | 63 |
GenBank mRNA | AF462610.1 | - | 2 - 3 | 192 | 63 |
AF537208.1 | - | 2 - 3 | 285 | 63 | |
BC087594.1 | - | - | 234 | 63 |