The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172047.1 | NP_742044.1 | 1 - 2 | 235 | 56 |
GenBank mRNA | AY049022.1 | - | 1 - 2 | 235 | 56 |