The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173291.1 | NP_775413.1 | 7 - 8 | 961 | 117 |
GenBank mRNA | AY649838.1 | - | 7 - 8 | 972 | 117 |
U56822.1 | - | 7 - 8 | 961 | 117 |