The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135698.1 | NP_001129170.1 | 1 - 2 | 129 | 165 |
GenBank mRNA | CB582520.1 | - | 1 - 2 | 141 | 165 |
FQ229460.1 | - | 1 - 2 | 118 | 165 | |
FQ229697.1 | - | 1 - 2 | 118 | 165 |