Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcolce
Entrez Gene ID
29569
Gene Name
procollagen C-endopeptidase enhancer
Gene Aliases
PCPE
Chromosome Location
Chr.12: 22153995 - 22160311 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019237.1 NP_062110.1 1 - 2 155 76
GenBank mRNA AB008534.1 - 1 - 2 142 76
AF016503.1 - 1 - 2 149 76
BC086572.1 - 1 - 2 136 76
FQ209381.1 - 1 - 2 191 76
U94710.1 - 1 - 2 155 76

View Full Product Details