Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hpx
Entrez Gene ID
58917
Gene Name
hemopexin
Gene Aliases
Hpxn
Chromosome Location
Chr.1: 170423558 - 170431066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053318.1 NP_445770.1 3 - 4 235 67
GenBank mRNA BC091137.1 - 3 - 4 226 67
FQ210231.1 - 3 - 4 236 67
FQ211176.1 - 3 - 4 236 67
FQ218633.1 - 3 - 4 234 67
FQ218971.1 - 3 - 4 228 67
FQ219309.1 - 3 - 4 234 67

View Full Product Details