Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pglyrp1
Entrez Gene ID
84387
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
Pglyrp, Pgrp
Chromosome Location
Chr.1: 79790879 - 79796064 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053373.1 NP_445825.1 1 - 2 266 80
GenBank mRNA AF154114.1 - 1 - 2 266 80

View Full Product Details