The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053472.1 | NP_445924.1 | 4 - 5 | 397 | 82 |
XM_006235280.2 | XP_006235342.1 | 4 - 5 | 752 | 82 | |
GenBank mRNA | AF255347.1 | - | 4 - 5 | 397 | 82 |