Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aldh9a1
Entrez Gene ID
64040
Gene Name
aldehyde dehydrogenase 9 family, member A1
Gene Aliases
Tmaba-dh
Chromosome Location
Chr.13: 85580828 - 85597497 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022273.2 NP_071609.2 10 - 11 1457 59
GenBank mRNA AF170918.1 - 10 - 11 1444 59
BC074019.1 - 10 - 11 1457 59

View Full Product Details