Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1c14
Entrez Gene ID
191574
Gene Name
aldo-keto reductase family 1, member C14
Gene Aliases
Akr1c9
Chromosome Location
Chr.17: 69806095 - 69827110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138547.2 NP_612556.1 7 - 8 894 69
GenBank mRNA BC091123.1 - 7 - 8 894 69
D17310.1 - 7 - 8 1082 69
M64393.1 - 7 - 8 893 69

View Full Product Details