Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a3
Entrez Gene ID
266730
Gene Name
solute carrier family 17 (organic anion transporter), member 3
Gene Aliases
Rnpt4
Chromosome Location
Chr.17: 43518760 - 43545159 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153622.2 NP_705886.1 5 - 6 702 67
XM_006253941.2 XP_006254003.1 8 - 9 1355 67
XM_006253942.2 XP_006254004.1 8 - 9 1357 67
XM_006253944.2 XP_006254006.1 8 - 9 1344 67
XM_006253946.2 XP_006254008.1 7 - 8 1231 67
XM_006253947.2 XP_006254009.1 8 - 9 1361 67
XM_006253948.2 XP_006254010.1 3 - 4 1232 67
XM_006253949.2 XP_006254011.1 6 - 7 1114 67
XM_006253950.2 XP_006254012.1 7 - 8 1124 67
XM_006253951.2 XP_006254013.1 6 - 7 998 67
XM_008771623.1 XP_008769845.1 9 - 10 1432 67
XM_008771624.1 XP_008769846.1 9 - 10 1420 67
XM_008771625.1 XP_008769847.1 7 - 8 1190 67
GenBank mRNA AB025223.1 - 5 - 6 726 67
BC078710.1 - 5 - 6 620 67

View Full Product Details