Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pah
Entrez Gene ID
24616
Gene Name
phenylalanine hydroxylase
Gene Aliases
-
Chromosome Location
Chr.7: 28066639 - 28129772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012619.2 NP_036751.2 5 - 6 598 77
XM_008765215.1 XP_008763437.1 3 - 4 538 77
GenBank mRNA BC078881.1 - 5 - 6 598 77
FQ219397.1 - 1 - 2 34 77
M12337.1 - 5 - 6 583 77

View Full Product Details