The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031138.2 | NP_112400.1 | 4 - 5 | 346 | 94 |
XM_006246343.2 | XP_006246405.1 | 5 - 6 | 371 | 94 | |
GenBank mRNA | BC070946.1 | - | 4 - 5 | 339 | 94 |
DN931313.1 | - | 4 - 5 | 376 | 94 | |
FQ216310.1 | - | 2 - 3 | 126 | 94 | |
FQ216742.1 | - | 1 - 2 | 65 | 94 | |
FQ222428.1 | - | 4 - 5 | 363 | 94 | |
FQ229894.1 | - | 4 - 5 | 357 | 94 | |
M62388.1 | - | 4 - 5 | 288 | 94 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008773893.1 | XP_008772115.1 | 5 - 6 | 370 | 94 |