Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ahcy
Entrez Gene ID
29443
Gene Name
adenosylhomocysteinase
Gene Aliases
-
Chromosome Location
Chr.3: 150587833 - 150603057 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017201.1 NP_058897.1 7 - 8 880 63
GenBank mRNA M15185.1 - 7 - 8 880 63

View Full Product Details