Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgals8
Entrez Gene ID
116641
Gene Name
lectin, galactoside-binding, soluble, 8
Gene Aliases
-
Chromosome Location
Chr.17: 66487239 - 66515306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053862.2 NP_446314.2 9 - 10 934 91
XM_006254166.2 XP_006254228.1 9 - 10 1200 91
XM_006254167.2 XP_006254229.1 9 - 10 946 91
XM_006254168.1 XP_006254230.1 9 - 10 973 91
GenBank mRNA BC072488.1 - 9 - 10 934 91
U09824.1 - 8 - 9 914 91

View Full Product Details