The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080583.1 | NP_542150.1 | 15 - 16 | 2171 | 100 |
XM_008767935.1 | XP_008766157.1 | 16 - 17 | 2928 | 100 | |
XM_008767936.1 | XP_008766158.1 | 15 - 16 | 2142 | 100 | |
XM_008767937.1 | XP_008766159.1 | 14 - 15 | 2057 | 100 | |
XM_008767938.1 | XP_008766160.1 | 14 - 15 | 2074 | 100 | |
GenBank mRNA | BC103481.1 | - | 16 - 17 | 2140 | 100 |
M77246.1 | - | 15 - 16 | 2171 | 100 |