Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm8
Entrez Gene ID
60590
Gene Name
glutamate receptor, metabotropic 8
Gene Aliases
Glur8, Gprc1h, Mglur8, mGluR8b, mGlur
Chromosome Location
Chr.4: 54474663 - 55399118 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022202.1 NP_071538.1 2 - 3 797 64
GenBank mRNA U63288.1 - 2 - 3 797 64

View Full Product Details