The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013028.1 | NP_037160.1 | 4 - 5 | 431 | 80 |
GenBank mRNA | AJ002259.1 | - | 4 - 5 | 431 | 80 |